LIF372154903 Life Chemicals Inc.

4-(1H-imidazol-2-yl)benzoic acid

  • MDL number MFCD00456057
  • Cas Number : 108035-45-6
  • Molecular Weight : 188.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart