LIF372154904 Life Chemicals Inc.

1-methyl-1H-benzimidazol-5-ol

  • MDL number MFCD00456136
  • Cas Number : 50591-22-5
  • Molecular Weight : 148.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart