LIF372154906 Life Chemicals Inc.

ethyl pyrazolo[1,5-a]pyrimidine-3-carboxylate

  • MDL number MFCD00847745
  • Cas Number : 115932-00-8
  • Molecular Weight : 191.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart