LIF372154908 Life Chemicals Inc.

5-methyl-1-(4-methyl-6-oxo-1,6-dihydropyrimidin-2-yl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD10005880
  • Cas Number : 1176632-40-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart