LIF372154913 Life Chemicals Inc.

(1-methyl-1H-imidazol-4-yl)methanol

  • MDL number MFCD06738905
  • Cas Number : 17289-25-7
  • Molecular Weight : 112.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form oil
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart