LIF372154924 Life Chemicals Inc.

[3-(morpholin-4-ylsulfonyl)benzyl]amine

  • MDL number MFCD09735163
  • Cas Number : 933989-32-3
  • Molecular Weight : 256.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart