LIF372154925 Life Chemicals Inc.

acenaphthylen-1(2H)-one

  • MDL number MFCD00156651
  • Cas Number : 2235-15-6
  • Molecular Weight : 168.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart