LIF372154929 Life Chemicals Inc.

1-(3-Methoxyphenoxy)-3-(methylamino)propan-2-ol

  • MDL number MFCD00776877
  • Cas Number : 66766-04-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart