LIF372154930 Life Chemicals Inc.

1-Amino-3-diethylamino-propan-2-ol

  • MDL number MFCD00025593

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form oil
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart