LIF372154932 Life Chemicals Inc.

[3-(4-Fluorobenzyl)-2,4-dioxo-1,3,8-triazaspiro[4.5]dec-8-yl]acetic acid

  • MDL number MFCD09965538
  • Cas Number : 1021117-39-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart