LIF372154941 Life Chemicals Inc.

1-(2-Isothiocyanatoethyl)-2-methyl-1H-indole

  • MDL number MFCD16652904
  • Cas Number : 1105192-89-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart