LIF372154970 Life Chemicals Inc.

3-Isocyanato-1-methyl-1H-indole

  • MDL number MFCD11588129
  • Cas Number : 124808-78-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart