LIF372154992 Life Chemicals Inc.

2-(2-aminothiazol-4-yl)-1-(4-phenylpiperazin-1-yl)ethan-1-one

  • MDL number MFCD09743137
  • Cas Number : 941869-37-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart