LIF372154993 Life Chemicals Inc.

2-[(anilinocarbonyl)amino]-4-methyl-1,3-thiazole-5-carboxylic acid

  • MDL number MFCD09743138
  • Cas Number : 941869-40-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart