LIF372155001 Life Chemicals Inc.

2-[(4-Fluorobenzoyl)amino]-1,3-thiazole-4-carboxylic acid

  • MDL number MFCD09743142
  • Cas Number : 941869-52-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart