LIF372155002 Life Chemicals Inc.

2-[(2-Fluorobenzoyl)amino]-1,3-thiazole-4-carboxylic acid

  • MDL number MFCD09743143
  • Cas Number : 941869-55-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart