LIF372155004 Life Chemicals Inc.

2-[(Anilinocarbonyl)amino]-1,3-thiazole-4-carboxylic acid

  • MDL number MFCD09743145
  • Cas Number : 941869-61-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart