LIF372155070 Life Chemicals Inc.

5-(2-methyl-1,3-thiazol-4-yl)-1,3,4-oxadiazol-2-amine

  • MDL number MFCD16652954
  • Cas Number : 1219827-53-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart