LIF372155131 Life Chemicals Inc.

5-(chloromethyl)-3-methyl-1,2,4-oxadiazole

  • MDL number MFCD08445357
  • Cas Number : 1192-81-0
  • Molecular Weight : 132.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart