LIF372155141 Life Chemicals Inc.

2-[(4,6-dimethylpyrimidin-2-yl)amino]-1,3-thiazole-4-carboxylic acid

  • MDL number MFCD21091740
  • Cas Number : 1248908-13-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart