LIF372155144 Life Chemicals Inc.

4-cyclopropyl-1,3-thiazol-2-amine

  • MDL number MFCD01571854
  • Cas Number : 324579-90-0
  • Molecular Weight : 140.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart