LIF372155152 Life Chemicals Inc.

{2-[(6-methylpyridin-2-yl)amino]-1,3-thiazol-4-yl}acetic acid

  • MDL number MFCD21091743
  • Cas Number : 1283109-29-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart