LIF372155161 Life Chemicals Inc.

{2-[(pyrimidin-2-ylcarbonyl)amino]-1,3-thiazol-4-yl}acetic acid

  • MDL number MFCD21091746
  • Cas Number : 1283109-10-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart