LIF372155174 Life Chemicals Inc.

2-methyl-4,5,6,7-tetrahydro-2H-indazol-3-amine

  • MDL number MFCD14706181
  • Cas Number : 26503-23-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart