LIF372155239 Life Chemicals Inc.

N-(2-furylmethyl)-6-methoxy-1,3-benzothiazol-2-amine

  • MDL number MFCD18447399
  • Cas Number : 16763-25-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart