LIF372155243 Life Chemicals Inc.

4,6-difluoro-2-(piperidin-4-yloxy)-1,3-benzothiazole

  • MDL number MFCD21091767
  • Cas Number : 1283108-95-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart