LIF372155250 Life Chemicals Inc.

5-(5,6-dihydro-1,4-dioxin-2-yl)-1,3,4-oxadiazol-2-amine

  • MDL number MFCD20228252
  • Cas Number : 1283108-68-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart