LIF372155252 Life Chemicals Inc.

1-(1,3-benzothiazol-2-yl)azetidine-3-carboxylic acid

  • MDL number MFCD20282427
  • Cas Number : 1283108-69-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart