LIF372155262 Life Chemicals Inc.

1-(4-fluoro-1,3-benzothiazol-2-yl)azetidine-3-carboxylic acid

  • MDL number MFCD21091779
  • Cas Number : 1283108-55-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart