LIF372155273 Life Chemicals Inc.

1-(6-fluoro-1,3-benzothiazol-2-yl)azetidine-3-carboxylic acid

  • MDL number MFCD21091790
  • Cas Number : 1283109-69-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart