LIF372155295 Life Chemicals Inc.

5-(4-bromophenyl)-3-methyl-1,2,4-oxadiazole

  • MDL number MFCD03069782
  • Cas Number : 71566-07-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart