LIF372155303 Life Chemicals Inc.

3-[(3-methyl-1,2,4-oxadiazol-5-yl)methyl]-1H-indole

  • MDL number MFCD16875137
  • Cas Number : 343966-69-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart