LIF372155312 Life Chemicals Inc.

6-(3-methyl-1,2,4-oxadiazol-5-yl)quinoxaline

  • MDL number MFCD21091826
  • Cas Number : 1283108-26-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart