LIF372155338 Life Chemicals Inc.

5-(3-methyl-1,2,4-oxadiazol-5-yl)-1H-indole

  • MDL number MFCD21091832
  • Cas Number : 1283108-48-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart