LIF372155349 Life Chemicals Inc.

tert-butyl 2-amino-3-cyano-4,7-dihydrothieno[2,3-c]pyridine-6(5H)-carboxylate

  • MDL number MFCD05664037
  • Cas Number : 150986-83-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart