LIF372155351 Life Chemicals Inc.

2-amino-N-methyl-1,3-oxazole-4-carboxamide

  • MDL number MFCD20039215
  • Cas Number : 1333960-70-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart