LIF372155357 Life Chemicals Inc.

N-(4-methoxy-1,3-benzothiazol-2-yl)-N-methylglycine

  • MDL number MFCD21091835
  • Cas Number : 1351597-05-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart