LIF372155360 Life Chemicals Inc.

N-methyl-N-[4-(methylthio)-1,3-benzothiazol-2-yl]glycine

  • MDL number MFCD21091838
  • Cas Number : 1352999-41-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart