LIF372155376 Life Chemicals Inc.

N-methyl-N-[6-(trifluoromethoxy)-1,3-benzothiazol-2-yl]glycine

  • MDL number MFCD21091854
  • Cas Number : 1353000-13-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart