LIF372155391 Life Chemicals Inc.

N-(4-chloro-1,3-benzothiazol-2-yl)-N-methylglycine

  • MDL number MFCD21091869
  • Cas Number : 1353000-10-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart