LIF372155410 Life Chemicals Inc.

4-(5-amino-1,3,4-oxadiazol-2-yl)-1-phenylpyrrolidin-2-one

  • MDL number MFCD15119564
  • Cas Number : 1030103-68-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart