LIF372155411 Life Chemicals Inc.

1,3,4-oxadiazol-2-amine

  • MDL number MFCD17677516
  • Cas Number : 3775-60-8
  • Molecular Weight : 85.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart