LIF372155413 Life Chemicals Inc.

[4-(1-benzofuran-2-yl)-1,3-thiazol-2-yl]acetonitrile

  • MDL number MFCD06380513
  • Cas Number : 851399-94-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart