LIF372155414 Life Chemicals Inc.

2-amino-6-ethyl-N-methyl-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-3-carboxamide

  • MDL number MFCD11651564
  • Cas Number : 1099624-25-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart