LIF372155418 Life Chemicals Inc.

tert-butyl 4-[(2-amino-1,3-thiazol-4-yl)methyl]piperazine-1-carboxylate

  • MDL number MFCD15147067
  • Cas Number : 856418-80-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart