LIF372155469 Life Chemicals Inc.

6-(methylthio)-N-(tetrahydrofuran-2-ylmethyl)-1,3-benzothiazol-2-amine

  • MDL number MFCD22587758
  • Cas Number : 1350988-96-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart