LIF372155470 Life Chemicals Inc.

4,6-dimethyl-N-(tetrahydrofuran-2-ylmethyl)-1,3-benzothiazol-2-amine

  • MDL number MFCD22587759
  • Cas Number : 1350989-21-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart