LIF372155480 Life Chemicals Inc.

2-hydrazino-4-(trifluoromethyl)-1,3-benzothiazole

  • MDL number MFCD08459018
  • Cas Number : 898748-11-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart