LIF372155485 Life Chemicals Inc.

5-(2-methylphenyl)-1,3,4-oxadiazole-2-thiol

  • MDL number MFCD00992514
  • Cas Number : 2503-66-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart