LIF372155489 Life Chemicals Inc.

5-(4-fluorophenyl)-1,3,4-oxadiazole-2-thiol

  • MDL number MFCD02670392
  • Cas Number : 41421-13-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart